Gene Transcripts

Gene Symbol
Aven
Entrez Gene ID
311299
Gene Name
apoptosis, caspase activation inhibitor
Gene Aliases
Chromosome Location
Chr. 3: 103980619 - 104117193 on Build Rnor_6.0
UniGene ID
Rn.145049
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107757.1 NP_001101227.1 5 702
XM_006234707.2 XP_006234769.1 6 1076
XM_008762093.1 XP_008760315.1 6 1076

View Full Product Details