Gene Transcripts

Gene Symbol
LOC316124
Entrez Gene ID
316124
Gene Name
similar to gonadotropin-regulated long chain acyl-CoA synthetase
Gene Aliases
Chromosome Location
Chr. 9: 10158745 - 10188013 on Build Rnor_6.0
UniGene ID
Rn.159567
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134566.1 NP_001128038.1 11 1717
XM_006244323.2 XP_006244385.1 10 1746

View Full Product Details