Gene Transcripts

Gene Symbol
Lin52
Entrez Gene ID
362763
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
RGD1306798
Chromosome Location
Chr. 6: 108167247 - 108264445 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006240356.2 XP_006240418.1 2, 3 563
XM_006240358.2 XP_006240420.1 2, 3 389
XM_008764889.1 XP_008763111.1 2, 3 386
XM_008764890.1 XP_008763112.1 2, 3 379
XM_343089.7 XP_343090.4 2, 3 375

View Full Product Details