Gene Transcripts

Gene Symbol
Olr24
Entrez Gene ID
404898
Gene Name
olfactory receptor 24
Gene Aliases
Chromosome Location
Chr. 1: 150100236 - 150101177 on Build Rnor_6.0
UniGene ID
Rn.141946
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000693.1 NP_001000693.1 1 113

View Full Product Details