Gene Transcripts

Gene Symbol
Smc1a
Entrez Gene ID
63996
Gene Name
structural maintenance of chromosomes 1A
Gene Aliases
SB1.8, SMC-1A, Smc1l1
Chromosome Location
Chr. X: 21710976 - 21755708 on Build Rnor_6.0
UniGene ID
Rn.11763
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031683.1 NP_113871.1 6, 7 1158
GenBank mRNA AJ005113.1 - 6, 7 1158

View Full Product Details