Gene Transcripts

Gene Symbol
Pllp
Entrez Gene ID
64364
Gene Name
plasmolipin
Gene Aliases
Tm4sf11
Chromosome Location
Chr. 19: 10731855 - 10752641 on Build Rnor_6.0
UniGene ID
Rn.1721
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022533.1 NP_071978.1 2, 3 524
GenBank mRNA BC078823.1 - 2, 3 404
U13617.1 - 2, 3 297
Z49858.1 - 2, 3 524

View Full Product Details