Gene Transcripts

Gene Symbol
Pou4f1
Entrez Gene ID
114503
Gene Name
POU class 4 homeobox 1
Gene Aliases
Brn3a, brn-3A
Chromosome Location
Chr. 15: 88618255 - 88622575 on Build Rnor_6.0
UniGene ID
Rn.232568
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770931.1 XP_008769153.1 2 1336
GenBank mRNA AF390075.1 - 1 191

View Full Product Details