Gene Transcripts

Gene Symbol
Semg1
Entrez Gene ID
25013
Gene Name
semenogelin I
Gene Aliases
RATSVPIIA, SVPIIA, SVS2P, Svp1, Svs2, Svs2p2
Chromosome Location
Chr. 3: 160627609 - 160630457 on Build Rnor_6.0
UniGene ID
Rn.34331
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012710.2 NP_036842.2 2 207
GenBank mRNA BC061977.1 - 2 207

View Full Product Details