Gene Transcripts

Gene Symbol
Dnph1
Entrez Gene ID
171047
Gene Name
2'-deoxynucleoside 5'-phosphate N-hydrolase 1
Gene Aliases
C6orf108, RGD620382, Rcl
Chromosome Location
Chr. 9: 16845764 - 16848514 on Build Rnor_6.0
UniGene ID
Rn.6997
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133525.1 NP_598209.1 3 331
GenBank mRNA U82591.1 - 3 331

View Full Product Details