Gene Transcripts

Gene Symbol
Adh1
Entrez Gene ID
24172
Gene Name
alcohol dehydrogenase 1 (class I)
Gene Aliases
Adh, Adh1a, Adh1c
Chromosome Location
Chr. 2: 243550657 - 243562243 on Build Rnor_6.0
UniGene ID
Rn.40222
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA FQ209590.1 - 6, 7 894
FQ218571.1 - 6, 7 896
FQ218640.1 - 1, 2 216
M15327.1 - 6, 7 867

View Full Product Details