Gene Transcripts

Gene Symbol
RT1-S2
Entrez Gene ID
24994
Gene Name
RT1 class Ib, locus S2
Gene Aliases
RT1.S1
Chromosome Location
Chr. 20: 3153057 - 3165495 on Build Rnor_6.0
UniGene ID
Rn.222446
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008857.1 NP_001008857.1 1, 2 49
XM_008772705.1 XP_008770927.1 1, 2 2644

View Full Product Details