Gene Transcripts

Gene Symbol
Drgx
Entrez Gene ID
252880
Gene Name
dorsal root ganglia homeobox
Gene Aliases
Drg11, Prrxl1
Chromosome Location
Chr. 16: 8823872 - 8853430 on Build Rnor_6.0
UniGene ID
Rn.10189
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145767.1 NP_665710.1 7 942
GenBank mRNA U29174.1 - 7 942

View Full Product Details