Gene Transcripts

Gene Symbol
Ntf4
Entrez Gene ID
25730
Gene Name
neurotrophin 4
Gene Aliases
NT4P, Ntf5
Chromosome Location
Chr. 1: 101405314 - 101408145 on Build Rnor_6.0
UniGene ID
Rn.44225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013184.3 NP_037316.2 3 741
GenBank mRNA S69323.1 - 3 741

View Full Product Details