Gene Transcripts

Gene Symbol
Vom2r53
Entrez Gene ID
286915
Gene Name
vomeronasal 2 receptor, 53
Gene Aliases
RGD1310577
Chromosome Location
Chr. 7: 13151592 - 13184258 on Build Rnor_6.0
UniGene ID
Rn.10806
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173131.2 NP_775154.1 3 1300
GenBank mRNA AF016178.1 - 3 1300

View Full Product Details