Gene Transcripts

Gene Symbol
Parl
Entrez Gene ID
287979
Gene Name
presenilin associated, rhomboid-like
Gene Aliases
Psarl
Chromosome Location
Chr. 11: 84517368 - 84544463 on Build Rnor_6.0
UniGene ID
Rn.7053
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035249.1 NP_001030326.1 9 1005
GenBank mRNA BC088226.1 - 9 923
BC105907.1 - 9 1005
BC126098.1 - 9 1037
BC158564.1 - 9 989 8374563
FQ214115.1 - 9 1000

View Full Product Details