Gene Transcripts

Gene Symbol
Vps37b
Entrez Gene ID
288659
Gene Name
vacuolar protein sorting 37 homolog B (S. cerevisiae)
Gene Aliases
RGD1304576
Chromosome Location
Chr. 12: 37984790 - 38023369 on Build Rnor_6.0
UniGene ID
Rn.144032
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105928.1 NP_001099398.1 4 445
XM_006249332.2 XP_006249394.1 3 928

View Full Product Details