Gene Transcripts

Gene Symbol
Olr1071
Entrez Gene ID
288823
Gene Name
olfactory receptor 1071
Gene Aliases
ratchr7-8424998-8424057_ORF
Chromosome Location
Chr. 7: 9591059 - 9592000 on Build Rnor_6.0
UniGene ID
Rn.138131
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000063.1 NP_001000063.1 1 576

View Full Product Details