Gene Transcripts

Gene Symbol
Cmtm1
Entrez Gene ID
291823
Gene Name
CKLF-like MARVEL transmembrane domain containing 1
Gene Aliases
Chromosome Location
Chr. 19: 859947 - 870470 on Build Rnor_6.0
UniGene ID
Rn.163396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029914.1 NP_001025085.1 3 1020
XM_006255052.2 XP_006255114.1 2 911
GenBank mRNA BC098029.1 - 3 1020

View Full Product Details