Gene Transcripts

Gene Symbol
Ddx19a
Entrez Gene ID
292022
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a
Gene Aliases
Ddx19, ZD10B
Chromosome Location
Chr. 19: 43251510 - 43271866 on Build Rnor_6.0
UniGene ID
Rn.228949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005381.2 NP_001005381.2 2, 3 172
GenBank mRNA BC079094.1 - 2, 3 143
CB582878.1 - 2, 3 172

View Full Product Details