Gene Transcripts

Gene Symbol
RGD1311558
Entrez Gene ID
292139
Gene Name
similar to 4930506M07Rik protein
Gene Aliases
Shootin1
Chromosome Location
Chr. 1: 280129082 - 280233840 on Build Rnor_6.0
UniGene ID
Rn.16095
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760533.1 XP_008758755.1 12 1270
GenBank mRNA BC085922.1 - 14 1416
EF055488.1 - 15 1426

View Full Product Details