Gene Transcripts

Gene Symbol
Gzmm
Entrez Gene ID
29252
Gene Name
granzyme M (lymphocyte met-ase 1)
Gene Aliases
Chromosome Location
Chr. 7: 12893431 - 12899004 on Build Rnor_6.0
UniGene ID
Rn.9838
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057183.1 NP_476531.1 3 317
GenBank mRNA DV216119.1 - 3 317
FQ233430.1 - 3 344
L05175.1 - 3 260

View Full Product Details