Gene Transcripts

Gene Symbol
Vom2r46
Entrez Gene ID
295091
Gene Name
vomeronasal 2 receptor, 46
Gene Aliases
Chromosome Location
Chr. 2: 154729854 - 154763094 on Build Rnor_6.0
UniGene ID
Rn.107047
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099502.1 NP_001092972.1 163

View Full Product Details