Gene Transcripts

Gene Symbol
Sp5
Entrez Gene ID
296510
Gene Name
Sp5 transcription factor
Gene Aliases
Chromosome Location
Chr. 3: 56766475 - 56769277 on Build Rnor_6.0
UniGene ID
Rn.217785
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106552.1 NP_001100022.1 2 531

View Full Product Details