Gene Transcripts

Gene Symbol
Zfhx2
Entrez Gene ID
305888
Gene Name
zinc finger homeobox 2
Gene Aliases
ZFH
Chromosome Location
Chr. 15: 33719313 - 33752665 on Build Rnor_6.0
UniGene ID
Rn.161359
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098803.1 NP_001092273.1 2 2066
XM_008770677.1 XP_008768899.1 1 3841
XM_008770678.1 XP_008768900.1 1 3841

View Full Product Details