Gene Transcripts

Gene Symbol
Spire1
Entrez Gene ID
307348
Gene Name
spire-type actin nucleation factor 1
Gene Aliases
Chromosome Location
Chr. 18: 63226557 - 63357423 on Build Rnor_6.0
UniGene ID
Rn.98481
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107381.1 NP_001100851.1 10 1472
XM_006254860.2 XP_006254922.1 12 1744
XM_006254862.2 XP_006254924.1 11 1702
XM_006254863.2 XP_006254925.1 12 1743
XM_006254864.2 XP_006254926.1 11 1701

View Full Product Details