Gene Transcripts

Gene Symbol
Bend5
Entrez Gene ID
362564
Gene Name
BEN domain containing 5
Gene Aliases
Chromosome Location
Chr. 5: 131257127 - 131312335 on Build Rnor_6.0
UniGene ID
Rn.87596
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108672.1 NP_001102142.1 5, 6 1283

View Full Product Details