Gene Transcripts

Gene Symbol
Bzw1
Entrez Gene ID
363232
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
Chromosome Location
Chr. 9: 65279675 - 65293471 on Build Rnor_6.0
UniGene ID
Rn.234367
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198789.2 NP_942084.1 9 1108
XM_006245000.2 XP_006245062.2 9 1107
GenBank mRNA BC061580.1 - 9 1112

View Full Product Details