Gene Transcripts

Gene Symbol
Zfp787
Entrez Gene ID
365176
Gene Name
zinc finger protein 787
Gene Aliases
RGD1310536, Znf787
Chromosome Location
Chr. 1: 71416026 - 71447195 on Build Rnor_6.0
UniGene ID
Rn.23427
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108904.1 NP_001102374.1 2 953
XM_006228193.2 XP_006228255.1 3 1117

View Full Product Details