Gene Transcripts

Gene Symbol
LOC367117
Entrez Gene ID
367117
Gene Name
similar to RIKEN cDNA 2900055D03
Gene Aliases
LRRGT00187
Chromosome Location
Chr. 8: 86281899 - 86286991 on Build Rnor_6.0
UniGene ID
Rn.123639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047919.1 NP_001041384.1 3 542
GenBank mRNA AY539938.1 - 3 542

View Full Product Details