Gene Transcripts

Gene Symbol
Il18rap
Entrez Gene ID
373540
Gene Name
interleukin 18 receptor accessory protein
Gene Aliases
Chromosome Location
Chr. 9: 47225543 - 47255798 on Build Rnor_6.0
UniGene ID
Rn.105609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767060.1 XP_008765282.1 4 1310
GenBank mRNA AJ550893.1 - 2 327

View Full Product Details