Gene Transcripts

Gene Symbol
Olr586
Entrez Gene ID
404860
Gene Name
olfactory receptor 586
Gene Aliases
Chromosome Location
Chr. 3: 75906945 - 75907880 on Build Rnor_6.0
UniGene ID
Rn.142889
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000660.1 NP_001000660.1 1 906

View Full Product Details