Gene Transcripts

Gene Symbol
Vom1r26
Entrez Gene ID
494245
Gene Name
vomeronasal 1 receptor 26
Gene Aliases
V1rm1
Chromosome Location
Chr. 14: 62309299 - 62310204 on Build Rnor_6.0
UniGene ID
Rn.129440
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009525.1 NP_001009525.1 1 21
GenBank mRNA AY510372.1 - 1 21

View Full Product Details