Gene Transcripts

Gene Symbol
Ctla2a
Entrez Gene ID
498690
Gene Name
cytotoxic T lymphocyte-associated protein 2 alpha
Gene Aliases
RGD1565540
Chromosome Location
Chr. 17: 4169695 - 4194700 on Build Rnor_6.0
UniGene ID
Rn.7778
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109115.1 NP_001102585.1 2, 3 228
XM_006253525.2 XP_006253587.1 2, 3 371
GenBank mRNA FQ220025.1 - 2, 3 241
FQ220118.1 - 2, 3 241
FQ220143.1 - 1, 2 469
FQ223251.1 - 2, 3 241
FQ229997.1 - 2, 3 172

View Full Product Details