Gene Transcripts

Gene Symbol
RGD1563217
Entrez Gene ID
499265
Gene Name
similar to RIKEN cDNA 4930451I11
Gene Aliases
Chromosome Location
Chr. 1: 198266108 - 198268065 on Build Rnor_6.0
UniGene ID
Rn.125914
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109155.1 NP_001102625.1 2 210
XM_006230320.2 XP_006230382.1 2 388
XM_006230321.2 XP_006230383.1 2 167

View Full Product Details