Gene Transcripts

Gene Symbol
Sit1
Entrez Gene ID
500449
Gene Name
signaling threshold regulating transmembrane adaptor 1
Gene Aliases
Chromosome Location
Chr. 5: 58985912 - 58987538 on Build Rnor_6.0
UniGene ID
Rn.47894
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024344.1 NP_001019515.1 3, 4 361
GenBank mRNA BC088199.1 - 3, 4 361
FQ225852.1 - 2, 3 276
FQ233921.1 - 2, 3 276

View Full Product Details