Gene Transcripts

Gene Symbol
Serhl2
Entrez Gene ID
500911
Gene Name
serine hydrolase-like 2
Gene Aliases
RGD1563386
Chromosome Location
Chr. 7: 123963381 - 123990103 on Build Rnor_6.0
UniGene ID
Rn.208269
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130579.1 NP_001124051.1 11 897
XM_008765775.1 XP_008763997.1 11 883

Gene Symbol
Rrp7a
Entrez Gene ID
362967
Gene Name
ribosomal RNA processing 7 homolog A (S. cerevisiae)
Gene Aliases
Cgi-96, RGD1311547
Chromosome Location
Chr. 7: 123980251 - 123990139 on Build Rnor_6.0
UniGene ID
Rn.4204
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130568.1 NP_001124040.1 7 2287

View Full Product Details