Gene Transcripts

Gene Symbol
Cbl
Entrez Gene ID
500985
Gene Name
Cbl proto-oncogene, E3 ubiquitin protein ligase
Gene Aliases
RGD1561386
Chromosome Location
Chr. 8: 48481256 - 48564719 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006242965.2 XP_006243027.1 12 2062
XM_576396.6 XP_576396.3 13 2194

View Full Product Details