Gene Transcripts

Gene Symbol
Ninj2
Entrez Gene ID
59115
Gene Name
ninjurin 2
Gene Aliases
Chromosome Location
Chr. 4: 152631201 - 152733631 on Build Rnor_6.0
UniGene ID
Rn.162768
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021595.2 NP_067606.1 1, 2 150
GenBank mRNA AB040815.1 - 1, 2 151
AF250322.1 - 1, 2 151

View Full Product Details