Gene Transcripts

Gene Symbol
Fam104a
Entrez Gene ID
619573
Gene Name
family with sequence similarity 104, member A
Gene Aliases
Chromosome Location
Chr. 10: 102179478 - 102200420 on Build Rnor_6.0
UniGene ID
Rn.98587
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034958.2 NP_001030130.2 2, 3 311
GenBank mRNA BC087161.1 - 2, 3 153
BC105876.1 - 2, 3 182 7129874

View Full Product Details