Gene Transcripts

Gene Symbol
Otulin
Entrez Gene ID
100362554
Gene Name
OTU deubiquitinase with linear linkage specificity
Gene Aliases
Fam105b
Chromosome Location
Chr. 2: 80267724 - 80293230 on Build Rnor_6.0
UniGene ID
Rn.228998
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729016.4 7 1011
XM_006232088.2 XP_006232150.1 7 1008
GenBank mRNA BC158613.1 - 7 966

View Full Product Details