Gene Transcripts

Gene Symbol
Gcnt1
Entrez Gene ID
64043
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
core 2 GlcNAc-T
Chromosome Location
Chr. 1: 236547734 - 236585037 on Build Rnor_6.0
UniGene ID
Rn.162516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022276.1 NP_071612.1 1 960
XM_006231147.2 XP_006231209.1 3 569
XM_006231150.2 XP_006231212.1 3 661
GenBank mRNA S79797.1 - 1 960

View Full Product Details