Gene Transcripts

Gene Symbol
Vbp1
Entrez Gene ID
681825
Gene Name
von Hippel-Lindau binding protein 1
Gene Aliases
Chromosome Location
Chr. 1: 148425047 - 148443505 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229645.2 XP_006229707.1 4 508
XM_006229646.2 XP_006229708.1 4 523

View Full Product Details