Gene Transcripts

Gene Symbol
Nup43
Entrez Gene ID
683983
Gene Name
nucleoporin 43
Gene Aliases
Chromosome Location
Chr. 1: 1771462 - 1781554 on Build Rnor_6.0
UniGene ID
Rn.216533
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128191.1 NP_001121663.1 5 665
XM_006227622.2 XP_006227684.1 6 1002
GenBank mRNA BC158756.1 - 6 739

View Full Product Details