Gene Transcripts

Gene Symbol
Cog2
Entrez Gene ID
690961
Gene Name
component of oligomeric golgi complex 2
Gene Aliases
Chromosome Location
Chr. 19: 57286967 - 57319329 on Build Rnor_6.0
UniGene ID
Rn.146004
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110494.1 NP_001103964.1 10 1208
XM_006255816.2 XP_006255878.1 10 1226

View Full Product Details