Gene Transcripts

Gene Symbol
Fabp3
Entrez Gene ID
79131
Gene Name
fatty acid binding protein 3, muscle and heart
Gene Aliases
Chromosome Location
Chr. 5: 148528854 - 148535597 on Build Rnor_6.0
UniGene ID
Rn.32566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024162.1 NP_077076.1 2 151
GenBank mRNA FQ217465.1 - 2 155
J02773.1 - 2 151
M18034.1 - 2 138

View Full Product Details