Gene Transcripts

Gene Symbol
Hes5
Entrez Gene ID
79225
Gene Name
hes family bHLH transcription factor 5
Gene Aliases
Chromosome Location
Chr. 5: 172364421 - 172365188 on Build Rnor_6.0
UniGene ID
Rn.22422
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024383.1 NP_077359.1 3 321
GenBank mRNA D12516.1 - 3 321

View Full Product Details