Gene Transcripts

Gene Symbol
Synrg
Entrez Gene ID
84479
Gene Name
synergin, gamma
Gene Aliases
Ap1gbp1, Syng
Chromosome Location
Chr. 10: 71278698 - 71357791 on Build Rnor_6.0
UniGene ID
Rn.122513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053419.1 NP_445871.1 14 2970
GenBank mRNA EF121977.1 - 12 2433
EF121978.1 - 12 2433
EF121979.1 - 13 2667
EF121980.1 - 13 2667
EF121981.1 - 14 2970
EF121982.1 - 12 2433

View Full Product Details