Gene Transcripts

Gene Symbol
Angpt2
Entrez Gene ID
89805
Gene Name
angiopoietin 2
Gene Aliases
Agpt2, Ang-2
Chromosome Location
Chr. 16: 75966480 - 76016147 on Build Rnor_6.0
UniGene ID
Rn.138360
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134454.1 NP_604449.1 8 1369
GenBank mRNA AF030378.1 - 2 281
AF311728.1 - 2 281
BC161931.1 - 8 1369 9021835

View Full Product Details