Gene Transcripts

Gene Symbol
VPS9D1-AS1
Entrez Gene ID
100128881
Gene Name
VPS9D1 antisense RNA 1
Gene Aliases
FAISL, MYU
Chromosome Location
Chr.16: 89711856 - 89718165 on Build GRCh38
Uni Gene
Hs.666282
Interrogated Sequence Translated Protein
RefSeq NR_036480.1
GenBank mRNA AK096286.1 -
BC061904.1 -

View Full Product Details