Gene Transcripts

Gene Symbol
MAPK8IP3
Entrez Gene ID
23162
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
Gene Aliases
JIP-3, JIP3, JSAP1, SYD2, syd
Chromosome Location
Chr. 16: 1706183 - 1770317 on Build GRCh38
UniGene ID
Hs.207763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040439.1 NP_001035529.1 2 475
NM_001318852.1 NP_001305781.1 2 512
NM_015133.4 NP_055948.2 2 512
XM_005255190.2 XP_005255247.1 2 512
XM_006720869.2 XP_006720932.1 2 315
XM_011522429.2 XP_011520731.1 2 512
XM_011522430.2 XP_011520732.1 2 512
XM_011522431.2 XP_011520733.1 2 512
XM_011522432.2 XP_011520734.1 2 512
XM_017023079.1 XP_016878568.1 2 512
XM_017023080.1 XP_016878569.1 2 512
XM_017023081.1 XP_016878570.1 1 31
GenBank mRNA AB028989.2 - 2 475
AB621811.1 - 2 500
AK024437.1 - 2 454
BC137123.1 - 2 427 9021120
BC137124.1 - 2 427 9021121
BC144486.1 - 2 427 9053013
BC150266.1 - 2 475 9007222
BY795428.2 - 2 235
CR749696.1 - 2 315
DA135446.1 - 2 512

View Full Product Details